Få mere at vide
Diethanolamine Fusidate, TRC
Artikelnummer. 30010184
Skift visning
Mængde:
100 mg
250 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30010184 2.5 g 2.5g
17984295 250 mg -
17983505 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010184 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4418802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16391-75-6
Molekylær formel C31 H48 O6 . C4 H11 N O2
Molekylvægt (g/mol) 621.84
Synonym 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, (3α,4α,8α,9β,11α,13α,14β,16β,17Z)-, compd. with 2,2'-iminobis[ethanol] (1:1), 29-Nor-8α,9β,13α,14β-dammara-17(20),24-dien-21-oic acid, 3α,11α,16β-trihydroxy-, 16-acetate, compd. with 2,2'-iminodiethanol (1:1), (Z)- (8CI), Ethanol, 2,2'-iminobis-, (3α,4α,8α,9β,11α,13α,14β,16β,17Z)-16-(acetyloxy)-3,11-dihydroxy-29-nordammara-17(20),24-dien-21-oate (salt) (9CI), Diethanolamine fusidate, Diethanolamine ent-(17Z)-16α-(acetyloxy)-3β,11β-dihydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dien-21-oate
IUPAC navn (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid;2-(2-hydroxyethylamino)ethanol
SMIL C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C.OCCNCCO
Tekniske data
Kemisk navn eller materiale Diethanolamine Fusidate
Renhedsgrad noter HPLC
Mængde 2.5 g
InChI formel InChI=1S/C31H48O6.C4H11NO2/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;6-3-1-5-2-4-7/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);5-7H,1-4H2/b26-20-;/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m0./s1
Formel vægt 621.4241
Procent renhed >95%