missing translation for 'onlineSavingsMsg'
Få mere at vide

Diethanolamine Fusidate, Mikromol™

Artikelnummer. 17933876
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17933876 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17933876 Leverandør Mikromol Leverandørnr. MM3832.000100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diethanolamine Fusidate
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Diethanolamine Fusidate
CAS 16391-75-6
Analyt- eller komponentnavne Diethanolamine Fusidate
Forsendelsestilstand Room Temperature
Molekylær formel C31 H48 O6 . C4 H11 N O2
Certificeringer/compliance ISO 17025
Synonym (3,4α,8α,9β,11α,13α,14β,16β,17Z)-16-(Acetyloxy)-3,11-dihydroxy-29-nordammara-17(20),24-dien-21-oic Acid compd. with 2,2'-Iminobis[ethanol]
InChI formel InChI=1S/C31H48O6.C4H11NO2/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;6-3-1-5-2-4-7/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);5-7H,1-4H2/b26-20-;/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m0./s1
SMIL C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C.OCCNCCO
IUPAC navn (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid;2-(2-hydroxyethylamino)ethanol
Molekylvægt (g/mol) 621.84
Mængde 100 mg
Formel vægt 621.4241 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.