Learn More
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Catalog Number p-7077325
Click to view available options
Quantity:
250 g
1 kg
111-42-2
C4H11NO2
105.14
MFCD00002843
ZBCBWPMODOFKDW-UHFFFAOYSA-N
diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
8113
CHEBI:28123
2-(2-hydroxyethylamino)ethanol
OCCNCCO
111-42-2
C4H11NO2
105.14
MFCD00002843
ZBCBWPMODOFKDW-UHFFFAOYSA-N
diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
8113
CHEBI:28123
2-(2-hydroxyethylamino)ethanol
OCCNCCO
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 27.0°C to 30.0°C
Density 1.0955g/mL
Boiling Point 268.0°C
Flash Point 179°C
Linear Formula (HOCH2CH2)2NH
Refractive Index 1.4750 to 1.4790
Quantity 250 g
Merck Index 15,3118
Specific Gravity 1.0955
Solubility Information Solubility in water: 954g/L (20°C).
Viscosity 352 mPa.s (30°C)
Formula Weight 105.14
Percent Purity ≥98.5% (GC) 1% max. (2−Aminoethanol) (GC) 1.0% max. (Triethanolamine) (GC)
Grade ACS Reagent
Chemical Name or Material Diethanolamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.