Få mere at vide
Didesmethyl Zolmitriptan (90%), TRC
Artikelnummer. 30133829
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133829 25 mg
30003410 5 mg
30004948 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133829 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44165025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 139264-15-6
Molekylær formel C14 H17 N3 O2
Molekylvægt (g/mol) 259.3
Synonym 2-Oxazolidinone, 4-[[3-(2-aminoethyl)-1H-indol-5-yl]methyl]-, (4S)-, 2-Oxazolidinone, 4-[[3-(2-aminoethyl)-1H-indol-5-yl]methyl]-, (S)-, (4S)-4-[[3-(2-Aminoethyl)-1H-indol-5-yl]methyl]-2-oxazolidinone, (S)-4-[[3-(2-Aminoethyl)-1H-indol-5-yl]methyl]oxazolidin-2-one, (4S)-4-[[3-(2-Aminoethyl)-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one
IUPAC navn (4S)-4-[[3-(2-aminoethyl)-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one
SMIL NCCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12
Tekniske data
Kemisk navn eller materiale Didesmethyl Zolmitriptan
Mængde 25 mg
InChI formel InChI=1S/C14H17N3O2/c15-4-3-10-7-16-13-2-1-9(6-12(10)13)5-11-8-19-14(18)17-11/h1-2,6-7,11,16H,3-5,8,15H2,(H,17,18)/t11-/m0/s1
Formel vægt 259.1321