Få mere at vide
Didesmethyl Cariprazine, TRC
Artikelnummer. 30009376
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30009376 2.5 mg 2.5mg
30005535 25 mg -
30003927 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009376 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4416632.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 839712-25-3
Molekylær formel C19H28Cl2N4O
Molekylvægt (g/mol) 399.36
Synonym N-[trans-4-[2-[4-(2,3-Dichlorophenyl)-1-piperazinyl]ethyl]cyclohexyl]urea
SMIL ClC(C(Cl)=CC=C1)=C1N(CC2)CCN2CC[C@H]3CC[C@H](NC(N)=O)CC3
Tekniske data
Kemisk navn eller materiale Didesmethyl Cariprazine
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C19H28Cl2N4O/c20-16-2-1-3-17(18(16)21)25-12-10-24(11-13-25)9-8-14-4-6-15(7-5-14)23-19(22)26/h1-3,14-15H,4-13H2,(H3,22,23,26)
Formel vægt 398.164
Procent renhed >95%