missing translation for 'onlineSavingsMsg'
Få mere at vide

Didesethyl Chloroquine Hydroxyacetamide, TRC

Artikelnummer. 30388645
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30388645 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30388645 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44098010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Didesethyl Chloroquine Hydroxyacetamide
CAS 1159977-30-6
Molekylær formel C16 H20 Cl N3 O2
Mængde 10 mg
Synonym 4-(4'-Glycolamidyl-1'-methyl-3,4-butylamino)-7-chloroquinoline
InChI formel InChI=1S/C16H20ClN3O2/c1-11(3-2-7-19-16(22)10-21)20-14-6-8-18-15-9-12(17)4-5-13(14)15/h4-6,8-9,11,21H,2-3,7,10H2,1H3,(H,18,20)(H,19,22)
SMIL CC(CCCNC(=O)CO)Nc1ccnc2cc(Cl)ccc12
IUPAC navn N-[4-[(7-chloro-4-quinolyl)amino]pentyl]-2-hydroxy-acetamide
Molekylvægt (g/mol) 321.8
Formel vægt 321.1244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.