Få mere at vide
Didesethyl Chloroquine-D4, TRC
Artikelnummer. 30379235
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30379235 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379235 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4409621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Didesethyl Chloroquine-d4
CAS 1215797-41-3
Renhedsgrad noter HPLC
Molekylær formel C14H14D4ClN3
Mængde 1 mg
Synonym Didesethyl Chloroquine D4
InChI formel InChI=1S/C14H18ClN3/c1-10(3-2-7-16)18-13-6-8-17-14-9-11(15)4-5-12(13)14/h4-6,8-10H,2-3,7,16H2,1H3,(H,17,18)/i2D2,7D2
SMIL [2H]C([2H])(N)C([2H])([2H])CC(C)Nc1ccnc2cc(Cl)ccc12
Molekylvægt (g/mol) 267.79
Formel vægt 267.144
Procent renhed >95