Få mere at vide
Didemethyl Citalopram-d6 Hydrobromide, TRC
Artikelnummer. 30375684
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30375684 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375684 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4394821MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Didemethyl Citalopram-d6 Hydrobromide
CAS 1286427-93-7
Molekylær formel C18H12D6BrFN2O
Mængde 1 mg
Synonym 1-(3-Aminopropyl-d6)-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile Hydrobromide (1:1)
InChI formel InChI=1S/C18H17FN2O.BrH/c19-16-5-3-15(4-6-16)18(8-1-9-20)17-7-2-13(11-21)10-14(17)12-22-18;/h2-7,10H,1,8-9,12,20H2;1H/i1D2,8D2,9D2;
SMIL Br.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC navn 1-(3-amino-1,1,2,2,3,3-hexadeuteriopropyl)-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
Molekylvægt (g/mol) 383.29
Formel vægt 382.0963