missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Dicycloverine Hydrochloride, Mikromol™
Description
Dicycloverine Hydrochloride
Specifications
Specifications
| Kemisk navn eller materiale | Dicycloverine Hydrochloride |
| CAS | 67-92-5 |
| Analyt- eller komponentnavne | Dicycloverine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H35 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | [1,1'-Bicyclohexyl]-1-carboxylic acid, 2-(diethylamino)ethyl ester, hydrochloride (9CI), [Bicyclohexyl]-1-carboxylic acid, 2-(diethylamino)ethyl ester hydrochloride, Atumin, Benacol, Bentomine, Bentyl hydrochloride, Di-Syntramine, Dicyclomine hydrochloride, Dicycloverin hydrochloride, Diocyl hydrochloride, Dyspas, Mamiesan, Merbentyl, Procyclomin, Wyovin hydrochloride, beta-Diethylaminoethyl 1-cyclohexylcyclohexanecarboxylate hydrochloride |
| InChI formel | InChI=1S/C19H35NO2.ClH/c1-3-20(4-2)15-16-22-18(21)19(13-9-6-10-14-19)17-11-7-5-8-12-17;/h17H,3-16H2,1-2H3;1H |
| SMIL | Cl.CCN(CC)CCOC(=O)C1(CCCCC1)C2CCCCC2 |
| IUPAC navn | 2-(diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?