Learn More
Dicyclopentadiene, Sigma-Aldrich™
Product Code. 15828910
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15828910 2.5 L 2.5 L
15808910 100 mL 100mL
15818910 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15828910 Supplier Sigma Aldrich Supplier No. 8030382500
 Restricted Item
Forventet forsendelse: 19-11-2026
Stabilized for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77-73-6
Specifications
Melting Point 11°C to 13°C
Density 0.98 g/cm3 at 20°C
Boiling Point 166°C/1013 hPa
MDL Number MFCD00078246
Quantity 2.5 L
Synonym Dicyclopentadiene, Cyclopentadiene dimer, 4,7-Methano-3a,4,7,7a-tetrahydroindene, Dicyclopentadiene
Molecular Weight (g/mol) 132.2
Grade Research