Få mere at vide
Dicyclohexyl Phthalate-d4, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phthalic acid, bis-cyclohexyl ester D4 |
| CAS | 358731-25-6 |
| Molekylær formel | C20 2H4 H22 O4 |
| Mængde | 10 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dicyclohexyl ester, Phthalic acid, bis-cyclohexyl ester D4, Dicyclohexyl phthalate-d4, Phthalic Acid, Bis-cyclohexyl Ester D4 (phenyl D4), 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dicyclohexyl ester, Phthalic acid, bis-cyclohexyl ester D4, Dicyclohexyl phthalate-d4 |
| InChI formel | InChI=1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2/i7D,8D,13D,14D |
| SMIL | [2H]c1c([2H])c([2H])c(C(=O)OC2CCCCC2)c(C(=O)OC3CCCCC3)c1[2H] |
| IUPAC navn | dicyclohexyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molekylvægt (g/mol) | 334.44 |
| Formel vægt | 334.2082 |