Learn More
Dicyclohexyl phthalate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 84-61-7 |
| Molecular Formula | C20H26O4 |
| MDL Number | MFCD00003849 |
| Quantity | 25 g |
| SMILES | O=C(OC1CCCCC1)C2=CC=CC=C2C(OC3CCCCC3)=O |
| Molecular Weight (g/mol) | 330.42 |
| Percent Purity | 95% |
| Chemical Name or Material | Dicyclohexyl phthalate |