Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dicyclanil |
| Emballage | Glass Bottle |
| CAS | 112636-83-6 |
| Farve | Beige |
| pH | 8.4 (10 g/L at 20°C) |
| Smeltepunkt | 255°C |
| Synonym | 4,6-Diamino-2-(cyclopropylamino)-5-pyrimidinecarbonitrile |
| Industry Type | Environmental Analysis |
| Molekylvægt (g/mol) | 190.21 |
| Mængde | 100 mg |
| Vis mere |