Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Kemisk navn eller materiale | Dicofol |
| CAS | 115-32-2 |
| CAS Max % | ≤ 100% |
| Beilstein | 1886299 |
| Farve | Colorless |
| Smeltepunkt | 78.5°C to 79.5°C |
| Kogepunkt | 193°C (0.13 hPa) |
| Synonym | 2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol |
| Molekylvægt (g/mol) | 370.5 g/mol |
| Mængde | 100 mg |