Få mere at vide
Dicofol-D8, TRC
Artikelnummer. 30377503
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30377503 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377503 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4367011MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dicofol-D8
CAS 2714413-20-2
Renhedsgrad noter HPLC
Molekylær formel C14HD8Cl5O
Mængde 1 mg
Synonym 4-Chloro-α-(4-chlorophenyl)-α-(trichloromethyl)-benzenemethanol-D8, 4,4'-Dichloro-α-(trichloromethyl)-benzhydrol-D8, 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethanol-D8, 2,2,2-Trichloro-1,1-di(4-chlorophenyl)ethanol-D8, Acarin-D8, Agravertin-D8, Chloretanol-D8, Dichlorokelthane-D8
InChI formel InChI=1S/C14H9Cl5O/c15-11-5-1-9(2-6-11)13(20,14(17,18)19)10-3-7-12(16)8-4-10/h1-8,20H/i1D,2D,3D,4D,5D,6D,7D,8D
SMIL OC(C(Cl)(Cl)Cl)(C1=C(C([2H])=C(C([2H])=C1[2H])Cl)[2H])C2=C(C([2H])=C(C([2H])=C2[2H])Cl)[2H]
IUPAC navn 2,2,2-trichloro-1,1-bis(4-chlorophenyl-2,3,5,6-d4)ethan-1-ol
Molekylvægt (g/mol) 378.52
Formel vægt 375.9598
Procent renhed >95