Få mere at vide
Dicloxacillin sodium salt monohydrate, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J61581.06
| Mængde | 5g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Kemiske identifikatorer
| 13412-64-1 | |
| 513.342 | |
| ASMFXKXNAWVTAY-MBGFYOAHSA-N | |
| (2S,5R,6R)-6-[[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate |
| C19H21Cl2N3NaO6S | |
| MFCD00056865 | |
| 131673926 | |
| [HH].CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] |
Tekniske data
| Dicloxacillin sodium salt | |
| C19H21Cl2N3NaO6S | |
| 5g | |
| Soluble in water | |
| [HH].CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] | |
| 513.342 | |
| 492.33 |
| 13412-64-1 | |
| MFCD00056865 | |
| 14,3084 | |
| ASMFXKXNAWVTAY-MBGFYOAHSA-N | |
| (2S,5R,6R)-6-[[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate | |
| 131673926 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.