Få mere at vide
Diclofop-methyl, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diclofop methyl |
| CAS | 51338-27-3 |
| Molekylær formel | C16 H14 Cl2 O4 |
| Mængde | 1 g |
| Synonym | (±)-Diclofop-methyl, Avenoxan, Dichlofop-methyl, Dichlorfop-methyl, Diclofop methyl ester, Diclofop-methyl, Diclosan, HOE 23408, Hoegrass, Hoelon, Illoxan, Iloxan, Methyl 2[4-(2,4-dichlorophenoxy)phenoxy]propionate, Methyl diclofop, Methyldichlofop |
| InChI formel | InChI=1S/C16H14Cl2O4/c1-10(16(19)20-2)21-12-4-6-13(7-5-12)22-15-8-3-11(17)9-14(15)18/h3-10H,1-2H3 |
| SMIL | COC(=O)C(C)Oc1ccc(Oc2ccc(Cl)cc2Cl)cc1 |
| IUPAC navn | methyl 2-[4-(2,4-dichlorophenoxy)phenoxy]propanoate |
| Molekylvægt (g/mol) | 341.19 |
| Formel vægt | 340.0269 |