Få mere at vide
Diclofop-methyl 100 μg/mL in Methanol, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diclofop methyl |
| CAS | 51338-27-3 |
| Analyt- eller komponentnavne | Diclofop methyl |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C16 H14 Cl2 O4 |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Propanoic acid, 2-[4-(2,4-dichlorophenoxy)phenoxy]-, methyl ester, (±)-Diclofop-methyl, Avenoxan, Dichlofop-methyl, Dichlorfop-methyl, Diclofop methyl ester, Diclofop-methyl, Diclosan, HOE 23408, Hoegrass, Hoelon, Illoxan, Iloxan, Methyl 2[4-(2,4-dichlorophenoxy)phenoxy]propionate, Methyl diclofop, Methyldichlofop |
| InChI formel | InChI=1S/C16H14Cl2O4/c1-10(16(19)20-2)21-12-4-6-13(7-5-12)22-15-8-3-11(17)9-14(15)18/h3-10H,1-2H3 |
| SMIL | O=C(OC)C(OC1=CC=C(OC2=CC=C(Cl)C=C2Cl)C=C1)C |
| Vis mere |