Få mere at vide
Diclofenac Sodium EP Impurity B, Mikromol™
Artikelnummer. 30052122
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052122 25 mg
17921946 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052122 Leverandør Mikromol Leverandørnr. MM0006.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diclofenac Sodium EP Impurity B
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(2,6-Dichlorophenyl)amino]benzaldehyde
CAS 22121-58-0
Analyt- eller komponentnavne 2-[(2,6-Dichlorophenyl)amino]benzaldehyde
Forsendelsestilstand Room Temperature
Molekylær formel C13 H9 Cl2 N O
Certificeringer/compliance ISO 17025
Synonym Diclofenac Impurity B, Diclofenac Sodium Imp. B (EP), Diclofenac Imp. B (EP), 2-[(2,6-Dichlorophenyl)amino]benzaldehyde, Anthranilaldehyde, N-(2,6-dichlorophenyl)- (8CI), 2-(2,6-Dichloroanilino)benzaldehyde, o-(2,6-Dichloroanilino)benzaldehyde, Diclofenac Sodium Impurity B, Diclofenac Impurity B
InChI formel InChI=1S/C13H9Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-8,16H
SMIL Clc1cccc(Cl)c1Nc2ccccc2C=O
IUPAC navn 2-(2,6-dichloroanilino)benzaldehyde
Molekylvægt (g/mol) 266.12
Mængde 25 mg
Formel vægt 265.0061 g/mol