Få mere at vide
Diclofenac Ethyl Ester, TRC
Artikelnummer. 30011911
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011911 250 mg
30003435 50 mg
30009918 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011911 Leverandør Toronto Research Chemicals Leverandørnr. TRCD436495250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15307-77-4
Molekylær formel C16 H15 Cl2 N O2
Molekylvægt (g/mol) 324.2
Synonym Ethyl [2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Aceclofenac Imp. C (EP), Ethyl Ester of Diclofenac
IUPAC navn ethyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
SMIL CCOC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
Tekniske data
Kemisk navn eller materiale Diclofenac Ethyl Ester
Mængde 250 mg
Renhedsgrad noter HPLC
InChI formel InChI=1S/C16H15Cl2NO2/c1-2-21-15(20)10-11-6-3-4-9-14(11)19-16-12(17)7-5-8-13(16)18/h3-9,19H,2,10H2,1H3
Formel vægt 323.048
Procent renhed >95%