Få mere at vide
Diclofenac-d4 (phenyl-d4-acetic), CDN
Artikelnummer. 30319800
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319800 0.005 g
30310852 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319800 Leverandør CDN Leverandørnr. CDND61830.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diclofenac-D4
CAS 153466-65-0
Molekylær formel C14 2H4 H7 Cl2 N O2
Mængde 0.005 g
Synonym 6-[(2,6-Dichlorophenyl)amino]-benzen-2,3,4,5-d4-acetic acid, Diclofenac D4 acid (phenyl D4), Diclofenac (phenyl D4)
InChI formel InChI=1 S/C14H11Cl2NO2/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19/h1-7,17 H,8H2,(H,18,19)/i1D,2 D,4 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(Nc2c(Cl)cccc2Cl)c(CC(=O)O)c1[2 H]
IUPAC navn 2-[2,3,4,5-tetradeuterio-6-(2,6-dichloroanilino)phenyl]acetic acid
Molekylvægt (g/mol) 300.17
Formel vægt 299.0418 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity