Få mere at vide
Diclofenac Amide, TRC
Artikelnummer. 17921935
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17921935 25 mg
17992035 250 mg
17984185 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17921935 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43644025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15362-40-0
Molekylær formel C14 H9 Cl2 N O
Molekylvægt (g/mol) 278.13
Synonym 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, Aceclofenac Imp. I (EP), Diclofenac Sodium Imp. A (EP), Diclofenac Related Compound A (USP)
IUPAC navn 1-(2,6-dichlorophenyl)-3H-indol-2-one
SMIL Clc1cccc(Cl)c1N2C(=O)Cc3ccccc23
Tekniske data
Kemisk navn eller materiale 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C14H9Cl2NO/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(17)18/h1-7H,8H2
Formel vægt 277.0061
Procent renhed >95%