Få mere at vide
Dichloroxylenol, LoGiCal
Artikelnummer. 30463070
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463070 10 mg
1 options
Artikelnummer. 30463070 Leverandør LoGiCal Leverandørnr. LGCFOR2546.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Dichloro-3,5-dimethylphenol
CAS 133-53-9
Analyt- eller komponentnavne 2,4-Dichloro-3,5-dimethylphenol
Forsendelsestilstand Room Temperature
Molekylær formel C8 H8 Cl2 O
Certificeringer/compliance ISO 9001
Synonym Phenol, 2,4-dichloro-3,5-dimethyl-, 3,5-Xylenol, 2,4-dichloro-, 2,4-Dichloro-3,5-dimethylphenol, 2,4-Dichloro-3,5-xylenol, DCMX, Decasept, Dichloroxylenol, Dixol, Hewsol, NSC 9774, Prinsyl
InChI formel InChI=1S/C8H8Cl2O/c1-4-3-6(11)8(10)5(2)7(4)9/h3,11H,1-2H3
Løsningstype Neat
SMIL Cc1cc(O)c(Cl)c(C)c1Cl
IUPAC navn 2,4-dichloro-3,5-dimethylphenol
Industry Type Clinical Testing
Molekylvægt (g/mol) 191.05
Mængde 10 mg
Formel vægt 189.9952 g/mol