Få mere at vide
Dichloroacetic Acid-OD, CDN
Artikelnummer. 30310844
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30310844 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310844 Leverandør CDN Leverandørnr. CDND06655G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dichloroacetic Acid-d1
CAS 82093-18-3
Molekylær formel C2 D H Cl2 O2
Mængde 5 g
Synonym Dichloro-acetic-d1 Acid
InChI formel InChI=1 S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i1D
SMIL [2 H]C(Cl)(Cl)C(=O)O
IUPAC navn 2,2-dichloro-2-deuterio-acetic acid
Molekylvægt (g/mol) 129.948
Formel vægt 128.9495 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity