Få mere at vide
Dichloroacetic Acid-d1, TRC
Artikelnummer. 30369877
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30369877 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369877 Leverandør Toronto Research Chemicals Leverandørnr. TRCD431829500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dichloroacetic Acid-d1
CAS 82093-18-3
Renhedsgrad noter GC
Molekylær formel C2 D H Cl2 O2
Mængde 500 mg
Synonym Dichloro-acetic-d1 Acid
InChI formel InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i1D
SMIL [2H]C(Cl)(Cl)C(=O)O
IUPAC navn 2,2-dichloro-2-deuterio-acetic acid
Molekylvægt (g/mol) 129.948
Formel vægt 128.9495
Procent renhed >95