Få mere at vide
Dibutylamine-d18, TRC
Artikelnummer. 30378364
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30378364 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378364 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4627372.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale di-n-Butylamine-d18
CAS 203578-32-9
Molekylær formel C8 D18 H N
Mængde 2.5 mg
Synonym 1-Butan-1,1,2,2,3,3,4,4,4-d9-amine, N-(butyl-d9)- (9CI), Di-n-butyl-d18-amine, Dibutylamine-d18, Dibutylamine-D18, N-(Butyl-1,1,2,2,3,3,4,4,4-d9)-1-butan-1,1,2,2,3,3,4,4,4-d9-amine
InChI formel InChI=1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,2,2,3,3,4,4,4-nonadeuterio-N-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)butan-1-amine
Molekylvægt (g/mol) 147.3541
Formel vægt 147.2647