missing translation for 'onlineSavingsMsg'
Få mere at vide

Dibutyl Phosphate-d18, TRC

Artikelnummer. 30378852
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378852 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378852 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4295225MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dibutyl Phosphate-d18
CAS 156213-21-7
Renhedsgrad noter HPLC
Molekylær formel C8HD18O4P
Mængde 5 mg
Synonym 1-Butan-1,1,2,2,3,3,4,4,4-d18-ol Hydrogen Phosphate, Butyl Phosphate, (BuO)2(HO)PO-d18, DBP-d18, DP 4-d18, DP 4 (coupling agent)-d18, Di-n-Butyl Phosphate-d18, Dibutyl Acid Phosphate-d18, Dibutyl Hydrogen Phosphate-d18, Dibutyl Phosphate-d18, HDBP-d18, NSC 203039-d18, NSC 22313-d18, Phosphoric Acid Dibutyl Ester-d18
InChI formel InChI=1S/C8H19O4P/c1-3-5-7-11-13(9,10)12-8-6-4-2/h3-8H2,1-2H3,(H,9,10)/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2
SMIL O=P(OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
Molekylvægt (g/mol) 228.32
Procent renhed >95
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.