Få mere at vide
Dibromomethane-d2, CDN
Artikelnummer. 30312600
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30312600 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312600 Leverandør CDN Leverandørnr. CDND01395G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dibromomethane-d2
CAS 22117-86-8
Molekylær formel C D2 Br2
Mængde 5 g
Synonym Dibromodideuteromethane, Dideuteriodibromomethane, Methylene dibromide-d2, Methylene-d2 bromide
InChI formel InChI=1 S/CH2Br2/c2-1-3/h1H2/i1D2
SMIL [2 H]C([2 H])(Br)Br
IUPAC navn dibromo(dideuterio)methane
Molekylvægt (g/mol) 175.85
Formel vægt 173.8649 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity