Få mere at vide
Dibenzylamine-d10, TRC
Artikelnummer. 30356540
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30356540 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30356540 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41750750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dibenzylamine-d10
CAS 923282-09-1
Molekylær formel C14 D10 H5 N
Mængde 50 mg
Synonym N-(Phenyl-2,3,4,5,6-d5-methyl)-benzene-2,3,4,5,6-d5-methanamine
InChI formel InChI=1S/C14H15N/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10,15H,11-12H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(CNCc2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC navn 1-(2,3,4,5,6-pentadeuteriophenyl)-N-[(2,3,4,5,6-pentadeuteriophenyl)methyl]methanamine
Molekylvægt (g/mol) 207.337
Formel vægt 207.183