missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Dibenzylphthalat-3,4,5,6-d4, CDN
Specifications
Specifications
| Kemisk navn eller materiale | Phthalic acid, bis-benzyl ester D4 |
| CAS | 1015854-62-2 |
| Molekylær formel | C22 2H4 H14 O4 |
| Mængde | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-bis(phenylmethyl) ester, Dibenzyl Phthalate-d4, Phthalic acid, bis-benzyl ester D4, Phthalic Acid Dibenzyl Ester (3,4,5,6)-D4 |
| InChI formel | InChI=1 S/C22H18O4/c23-21(25-15-17-9-3-1-4-10-17)19-13-7-8-14-20(19)22(24)26-16-18-11-5-2-6-12-18/h1-14 H,15-16H2/i7D,8 D,13 D,14 D |
| SMIL | [2 H]c1c([2 H])c([2 H])c(C(=O)OCc2ccccc2)c(C(=O)OCc3ccccc3)c1[2 H] |
| IUPAC navn | dibenzyl 3,4,5,6-tetradeuteriobenzen-1,2-dicarboxylat |
| Molekylvægt (g/mol) | 350.4 |
| Formel vægt | 350,1456 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?