missing translation for 'onlineSavingsMsg'
Få mere at vide

Dibenzyl Hydrogen Phosphate, TRC

Artikelnummer. 30361342
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361342 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30361342 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4177201G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dibenzyl Hydrogen Phosphate
CAS 538-60-3
Molekylær formel C14 H15 O3 P
Mængde 1 g
InChI formel InChI=1S/C14H15O3P/c15-18(16-11-13-7-3-1-4-8-13)17-12-14-9-5-2-6-10-14/h1-10,18H,11-12H2
SMIL O=P(OCc1ccccc1)OCc2ccccc2
IUPAC navn phenylmethoxyphosphonoyloxymethylbenzene
Molekylvægt (g/mol) 262.24
Formel vægt 262.0759
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.