Learn More
Alfa Aesar™ Dibenzothiophene-4-carboxaldehyde, 98%
Brand: Alfa Aesar™ H64515.06
Additional Details : CAS Number : 23985-81-1 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Dibenzothiophene-4-carboxaldehyde | |
212.27 | |
1567946 |
23985-81-1 | |
MFCD00961967 |
Specifications
Dibenzothiophene-4-carboxaldehyde | |
Powder | |
Pale yellow | |
Air Sensitive | |
MFCD00961967 |
23985-81-1 | |
98% | |
5g | |
212.27 | |
1567946 |