Få mere at vide
Dibenzothiophene-1,2,3,4,6,7,8,9-d8, TRC
Artikelnummer. 30359562
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30359562 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359562 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41737450MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dibenzothiophene D8
CAS 33262-29-2
Molekylær formel C12 2H8 S
Mængde 50 mg
Synonym Dibenzothiophene D8, Dibenzothiophene-1,2,3,4,6,7,8,9-d8 (ACI), Dibenzothiophene-d8 (8CI, 9CI)
InChI formel InChI=1S/C12H8S/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c2c(sc3c([2H])c([2H])c([2H])c([2H])c23)c1[2H]
IUPAC navn 1,2,3,4,6,7,8,9-octadeuteriodibenzothiophene
Molekylvægt (g/mol) 192.31
Formel vægt 192.0849