missing translation for 'onlineSavingsMsg'
Få mere at vide

Dibenzo[de,qr]naphthacene-9,14-dione, TRC

Artikelnummer. 30356858
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356858 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30356858 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4174951MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Kemisk navn eller materiale Dibenzo[de,qr]naphthacene-9,14-dione
CAS 169828-20-0
Molekylær formel C24H12O2
Mængde 1 mg
InChI formel InChI=1S/C24H12O2/c25-23-15-7-1-2-8-16(15)24(26)22-18-10-4-6-14-12-11-13-5-3-9-17(21(22)23)19(13)20(14)18/h1-12H
SMIL O=C1c2ccccc2C(=O)c3c1c4cccc5ccc6cccc3c6c45
Molekylvægt (g/mol) 332.35
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.