missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo-18-crown-6, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.
Specifications
Melting Point | 160°C to 164°C |
Boiling Point | 380°C to 384°C (679 mmHg) |
Quantity | 5 g |
Beilstein | 1162153 |
Merck Index | 14,2602 |
Solubility Information | Sparingly soluble in water. |
SMILES | C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1 |
Molecular Weight (g/mol) | 360.406 |
Formula Weight | 360.41 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'