Få mere at vide
Dibenzazepinone-d4 (Major), TRC
Artikelnummer. 30351270
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30351270 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351270 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4168925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dibenzazepinone-d4
CAS 1189706-86-2
Molekylær formel C14 2H4 H7 N
Mængde 5 mg
InChI formel InChI=1S/C14H11N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-10,15H/i1D,3D,5D,7D
SMIL [2H]c1c([2H])c([2H])c2C=Cc3ccccc3Nc2c1[2H]
IUPAC navn 1,2,3,4-tetradeuterio-11H-benzo[b][1]benzazepine
Molekylvægt (g/mol) 197.27
Formel vægt 197.1143