Få mere at vide
Diazoxide-d3, TRC
Artikelnummer. 30361800
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30361800 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361800 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4173081MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diazoxide-d3
CAS 1432063-51-8
Renhedsgrad noter HPLC
Molekylær formel C8H4D3ClN2O2S
Mængde 1 mg
Synonym 7-Chloro-3-methyl-d3-2H-1,2,4-benzothiadiazine 1,1-Dioxide, 3-Methyl-d3-7-chloro-1,2,4-benzothiadiazine 1,1-Dioxide, 7-Chloro-3-methyl-d3-2H-1,2,4-benzothiadiazine 1,1-Dioxide, Dizoxide-d3, Hyperstat-d3, Hypertonalum-d3, Mutabase-d3, NSC 64198-d3, NSC 76130-d3, Proglicem-d3, Proglycem-d3, SRG 95213-d3, Sch 6783-d3
InChI formel InChI=1S/C8H7ClN2O2S/c1-5-10-7-3-2-6(9)4-8(7)14(12,13)11-5/h2-4H,1H3,(H,10,11)/i1D3
SMIL O=S1(C2=CC(Cl)=CC=C2NC(C([2H])([2H])[2H])=N1)=O
Molekylvægt (g/mol) 233.69
Procent renhed >95