Få mere at vide
Diazaborine, TRC
Artikelnummer. 30367716
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367716 50 mg
30359957 25 mg
30350405 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367716 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41759350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22959-81-5
Molekylær formel C14H13BN2O3S
Molekylvægt (g/mol) 300.14
Synonym 1,2-Dihydro-1-hydroxy-2-(p-tolylsulfonyl)-2,3,1-benzodiazaborine, 1,2-Dihydro-1-hydroxy-2-(p-tosyl)-2,3,1-benzo[e]diazaborine, 1,2-Dihydro-1-hydroxy-2-[(4-methylphenyl)sulfonyl]-2,3,1-benzodiazaborine
SMIL Cc1ccc(cc1)S(=O)(=O)N2N=Cc3ccccc3B2O
Tekniske data
Kemisk navn eller materiale Diazaborine
Mængde 50 mg
InChI formel InChI=1S/C14H13BN2O3S/c1-11-6-8-13(9-7-11)21(19,20)17-15(18)14-5-3-2-4-12(14)10-16-17/h2-10,18H,1H3
Formel vægt 300.074