missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diatrizoic acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
Soluble in Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
Specifications
Color | White |
Quantity | 5 g |
Merck Index | 14,2992 |
Solubility Information | Soluble in Methanol. |
Formula Weight | 613.9 |
Physical Form | Powder |
Chemical Name or Material | Diatrizoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'