missing translation for 'onlineSavingsMsg'
Få mere at vide

Diamino Biotin, TRC

Artikelnummer. 30137879
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137879 250 mg
17940885 50 mg
17920895 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30137879 Leverandør Toronto Research Chemicals Leverandørnr. TRCD416235250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 22342-46-7
Molekylær formel C9 H18 N2 O2 S
Molekylvægt (g/mol) 218.32
Synonym (2S,3S,4R)-3,4-Diaminotetrahydro-2-thiophenepentanoic Acid, 2-Thiophenepentanoic acid, 3,4-diaminotetrahydro-, [2S-(2α,3α,4α)]-, 2-Thiophenevaleric acid, 3,4-diaminotetrahydro-, (2S,3S,4R)- (8CI), (2S,3S,4R)-3,4-Diaminotetrahydro-2-thiophenepentanoic acid, Diaminobiotin
IUPAC navn 5-[(2S,3S,4R)-3,4-diaminothiolan-2-yl]pentanoic acid
SMIL N[C@H]1CS[C@@H](CCCCC(=O)O)[C@H]1N

Specifications

Kemisk navn eller materiale Diaminobiotin
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C9H18N2O2S/c10-6-5-14-7(9(6)11)3-1-2-4-8(12)13/h6-7,9H,1-5,10-11H2,(H,12,13)/t6-,7-,9-/m0/s1
Formel vægt 218.1089
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.