missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl maleate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.
Solubility
Soluble in water 151.2 mg/L @ 25°C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.
Solubility
Soluble in water 151.2 mg/L @ 25°C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | -47°C |
Density | 1.072 |
Boiling Point | 109°C to 110°C (3 mmHg) |
Flash Point | 130°C (266°F) |
Refractive Index | 1.467 |
Quantity | 50 g |
UN Number | UN2810 |
Beilstein | 1725954 |
Solubility Information | Soluble in water 151.2mg/L @ 25°C. |
Formula Weight | 196.21 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'