Learn More
Diaion™ UBK550(Na), ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1.9 meq/ml on PS-DVB, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 046609.A1
645.87 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 1.00000kg
Product Code. 15494055
  • 968.77 DKK / 1kg
Estimated Shipment: 02-05-2024
to see stock.
Description
Description
Chromatographic oligosaccharide sugar separations, sugar alcohol separations
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
220μm Beads
8% cross-linked, strongly acidic gel type, 1.9meq/mL on PS-DVB
1 kg
Diaion™ UBK550 (Na), ion exchange fractionation resin
220μm
Product Suggestions
Product Suggestions
Videos
SDS Documents