Learn More
Diaion™ CR20, chelating resin, highly porous type, 0.4 mmole/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046781.A1
929.07 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
Transitional metal removal without alkali earth metal adsorption
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
300-1180μm Beads | |
Highly porous type, 0.4mmole/mL on poly(styrene-divinylbenzene) | |
300 to 1180μm | |
1 kg |
Diaion™ CR20, chelating resin | |
Odorless | |
Yellow |
RUO – Research Use Only