Få mere at vide
Diacetoxyscirpenol, TRC
Artikelnummer. 30353738
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30353738 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353738 Leverandør Toronto Research Chemicals Leverandørnr. TRCD3075505MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diacetoxyscirpenol
CAS 2270-40-8
Renhedsgrad noter HPLC
Molekylær formel C19 H26 O7
Mængde 5 mg
Synonym Anguidin (7CI), Trichothec-9-ene-3α,4β,15-triol, 12,13-epoxy-, 4,15-diacetate (8CI), 4,15-Di-O-acetylscirpenol, 4,15-Diacetoxyscirp-9-en-3-ol, 4,15-Diacetoxyscirpen-3-ol, 4,15-Diacetoxyscirpenol, 4β,15-Diacetoxy-12,13-epoxytrichothec-9-en-3α-ol, Anguidine, DAS, Diacetoxyscirpenol, NSC 141537, NSC 177378, Scirp-9-ene-3α,4β,15-triol, 4,15-diacetate, Scirpenetriol 4,15-diacetate
InChI formel InChI=1S/C19H26O7/c1-10-5-6-18(8-23-11(2)20)13(7-10)26-16-14(22)15(25-12(3)21)17(18,4)19(16)9-24-19/h7,13-16,22H,5-6,8-9H2,1-4H3/t13-,14-,15-,16-,17-,18-,19-/m1/s1
SMIL CC(=O)OC[C@]12CCC(=C[C@H]1O[C@@H]3[C@H](O)[C@@H](OC(=O)C)[C@@]2(C)[C@@]34CO4)C
Molekylvægt (g/mol) 366.41
Formel vægt 366.1679
Procent renhed >95