Få mere at vide
Diacetoxybutene, TRC
Artikelnummer. 30179978
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179978 100 mg
30179977 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179978 Leverandør Toronto Research Chemicals Leverandørnr. TRCD422425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18085-02-4
Molekylær formel C8 H12 O4
Molekylvægt (g/mol) 172.1785
Synonym 3-Butene-1,2-diol, diacetate (8CI,9CI), 1,2-Diacetoxy-3-butene, 1-Butene-3,4-diol diacetate, 1-Butene-3,4-diyl diacetate, 2-(Acetoxymethyl)-3-acetoxy-1-propene, 3,4-Diacetoxy-1-butene, 3,4-Diacetyloxy-1-butene, Diacetoxybutene, 3-Butene-1,2-diol, 1,2-diacetate
IUPAC navn 2-acetyloxybut-3-enyl acetate
SMIL CC(=O)OCC(OC(=O)C)C=C
Tekniske data
Kemisk navn eller materiale 2-(Acetoxymethyl)-3-acetoxy-1-propene
Mængde 100 mg
InChI formel InChI=1S/C8H12O4/c1-4-8(12-7(3)10)5-11-6(2)9/h4,8H,1,5H2,2-3H3
Formel vægt 172.0736