missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diacetone acrylamide, 99%, Thermo Scientific Chemicals
561.41 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.
Solubility
Soluble in water, methanol, benzene and tetrahydrofuran.
Notes
Store in cool place. Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.
Solubility
Soluble in water, methanol, benzene and tetrahydrofuran.
Notes
Store in cool place. Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Specifications
Melting Point | 54°C to 56°C |
Flash Point | >110°C (230°F) |
Quantity | 250 g |
Beilstein | 1928444 |
Sensitivity | Hygroscopic |
Merck Index | 14,2963 |
Solubility Information | Soluble in water,methanol,benzene and tetrahydrofuran. |
Formula Weight | 169.23 |
Percent Purity | 99% |
Chemical Name or Material | Diacetone acrylamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'