Få mere at vide
Diacetolol Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diacetolol Hydrochloride |
| CAS | 73899-76-0 |
| Analyt- eller komponentnavne | Diacetolol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H24 N2 O4 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Acebutolol Hydrochloride Imp. B (EP), Acebutolol Imp. B (EP), Acebutolol USP Related Compound B, Acebutolol USP RC B, N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetamide Hydrochloride, Acebutolol Hydrochloride Imp. B (EP) as Hydrochloride, Diacetolol Hydrochloride, Acebutolol Related Compound B as Hydrochloride, Acebutolol Hydrochloride Impurity B as Hydrochloride, Acebutolol Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C16H24N2O4.ClH/c1-10(2)17-8-14(21)9-22-16-6-5-13(18-12(4)20)7-15(16)11(3)19;/h5-7,10,14,17,21H,8-9H2,1-4H3,(H,18,20);1H |
| Løsningstype | Neat |
| SMIL | Cl.CC(C)NCC(O)COc1ccc(NC(=O)C)cc1C(=O)C |
| Vis mere |