Learn More
di-Sodium Tetraborate, Supelco™
Specifications
Specifications
| Melting Point | 742°C |
| CAS | 1330-43-4 |
| Quantity | 25 g |
| MDL Number | MFCD00081185 |
| Synonym | di-Sodium tetraborate, Borax, di-Sodium tetraborate |
| Molecular Weight (g/mol) | 201.22 |
| Grade | Research |
| Density | 2.37 g/cm3 at 20°C |
| Chemical Name or Material | Di-Sodium Tetraborate |