Learn More
di-Sodium hydrogen phosphate dodecahydrate, Supelco™
Specifications
Specifications
| Chemical Name or Material | di-Sodium hydrogen phosphate dodecahydrate |
| CAS | 10039-32-4 |
| Quantity | 25 kg |
| Density | 1.52 g/cm³ at 20°C |
| Molecular Formula | Na2HPO4 · 12H2O |
| Synonym | Sodium phosphate dibasic dodecahydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate |
| Molecular Weight (g/mol) | 358.14 |
| Grade | Not Available |