missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Di-natriumhydrogenorthophosphatdihydrat, ekstra rent, SLR, Fisher Chemical™
Shop alle Fisher Chemical produkter
Klik for at se tilgængelige muligheder
Mængde:
500 g
1 kg
5 kg
Emballage:
Plast pulver krukke
Plastfad
Pakningsstørrelse:
1kg
500g
5kg
Specifications
Specifications
| Smeltepunkt | 243°C |
| Farve | Hvid |
| pH | 8.7 to 9.4 |
| Fysisk form | Fast stof |
| Mængde | 500 g |
| Formel vægt | 177.99g/mol |
| Emballage | Plast pulver krukke |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction